Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
cis
-4-Hydroxy-L-proline
Synonym(s):
(2
S
,4
S
)-(−)-4-Hydroxy-2-pyrrolidinecarboxylic acid, CHP
Empirical Formula (Hill Notation):
C
5
H
9
NO
3
CAS Number:
618-27-9
Molecular Weight:
131.13
EC Number:
210-542-1
Products
Product Category
amino acids (1)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Optical Isomer
Amino Acid
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
H1637
cis
-4-Hydroxy-L-proline,
collagen synthesis inhibitor
Expand
View Pricing