Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chloroquine diphosphate salt
Synonym(s):
N
4
-(7-Chloro-4-quinolinyl)-
N
1
,
N
1
-dimethyl-1,4-pentanediamine diphosphate salt
Empirical Formula (Hill Notation):
C
18
H
26
ClN
3
· 2H
3
PO
4
CAS Number:
50-63-5
Molecular Weight:
515.86
EC Number:
200-055-2
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
C6628
Chloroquine diphosphate salt,
98.5-101.0% (EP), powder or crystals, anti-malarial drug
Expand
View Pricing