Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chloral hydrate
Synonym(s):
Trichloroacetaldehyde hydrate
Linear Formula:
Cl
3
CCH(OH)
2
CAS Number:
302-17-0
Molecular Weight:
165.40
EC Number:
206-117-5
Products
Product Category
building blocks (3)
bioactive small molecules (1)
Brand
Sigma-Aldrich (3)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Purity
Research Area
Quality Segment
Reagent Type
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
C8383
Chloral hydrate,
Expand
View Pricing
15307
Chloral hydrate,
≥99%
Expand
View Pricing
05-2660
Chloral hydrate,
SAJ first grade, ≥99.5%
Expand
View Pricing
05-2650
Chloral hydrate,
SAJ special grade, ≥99.7%
Expand
View Pricing