Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cerium(IV) sulfate solution
Synonym(s):
Ceric sulfate
Linear Formula:
Ce(SO
4
)
2
CAS Number:
13590-82-4
Molecular Weight:
332.24
Products
Brand
Supelco (1)
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Reaction Type
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
1.60338
Cerium(IV) sulfate solution,
c(Ce(SO₄)2 * 4 H₂O) = 0.1 mol/l (0.1 N), reag. USP, ready-to-use volumetric solution for titration, Titriplex
®
Expand
View Pricing