Cerium(III) acetylacetonate hydrate
Synonym(s):
Ce(acac)3, Cerium(III) 2,4-pentanedionate
Linear Formula:
Ce(C5H7O2)3 · xH2O
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing