Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Calcium L-lactate pentahydrate
Synonym(s):
L-Lactic acid calcium salt
Linear Formula:
[CH
3
CH(OH)COO]
2
Ca · 5H
2
O
CAS Number:
5743-47-5
Molecular Weight:
308.29
EC Number:
248-953-3
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Optical Isomer
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
21175
Calcium L-lactate pentahydrate,
tested according to Ph. Eur.
Expand
View Pricing
05-0810
Calcium L-lactate pentahydrate,
SAJ first grade, ≥97.0%
Expand
View Pricing