Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Caffeic acid
Synonym(s):
3,4-Dihydroxybenzeneacrylic acid, 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)-2-propenoic acid, Caffeic Acid
Linear Formula:
(HO)
2
C
6
H
3
CH=CHCO
2
H
CAS Number:
331-39-5
Molecular Weight:
180.16
EC Number:
206-361-2
Products
Product Category
bioactive small molecules (2)
acids (1)
antioxidants (1)
flavonoids (1)
natural products (1)
phenylpropanoids (1)
Show More
Brand
Sigma-Aldrich (2)
Supelco (1)
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Analyte Suitability
Research Area
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Natural (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
C0625
Caffeic acid,
≥98.0% (HPLC)
Expand
View Pricing
205546
Caffeic acid,
A natural dietary compound reported to have anti-carcinogenic and anti-inflammatory properties.
Expand
View Pricing
60018
Caffeic acid,
matrix substance for MALDI-MS, ≥99.0% (HPLC)
Expand
View Pricing