
Butyl parahydroxybenzoate impurity E

Synonym(s):
Isobutyl 4-hydroxybenzoate
Empirical Formula (Hill Notation):
C11H14O3
CAS Number:
Molecular Weight:
194.23
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing