Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Buffer, reference standard
Products
Product Category
buffers (3)
Brand
Sigma-Aldrich (1)
Supelco (1)
Color
Feature
pH Value
Physical Form
Quality Segment
Reference Material Type
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
B5020
Buffer, reference standard,
pH 4.00±0.01 (25 °C)
Expand
View Pricing
B4770
Buffer, reference standard,
pH 7.00±0.01 (25 °C)
Expand
View Pricing
B4895
Buffer, reference standard,
pH 10.00±0.01 (25 °C)
Expand
View Pricing