Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
insert strips for 24 x 6-well BRANDplates
®
Products
Product Category
inserts (12)
Manufacturer
Binding Type
Membrane Material
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
BR782870
BRAND
®
insert strips for 24 x 6-well BRANDplates
®
,
PET membrane, pore size 8 μm
Expand
View Pricing
BR782820
BRAND
®
insert strips for 24 x 6-well BRANDplates
®
,
PC membrane, pore size 1 μm
Expand
View Pricing
BR782810
BRAND
®
insert strips for 24 x 6-well BRANDplates
®
,
PET membrane, pore size 0.4 μm
Expand
View Pricing
BR782800
BRAND
®
insert strips for 24 x 6-well BRANDplates
®
,
PC membrane, pore size 0.4 μm
Expand
View Pricing
BR782840
BRAND
®
insert strips for 24 x 6-well BRANDplates
®
,
PC membrane, pore size 3 μm
Expand
View Pricing
BR782860
BRAND
®
insert strips for 24 x 6-well BRANDplates
®
,
PC membrane, pore size 8 μm
Expand
View Pricing
BR782821
BRAND
®
insert strips for 24 x 6-well BRANDplates
®
,
Inlet Opening System, PC membrane, pore size 1 μm
Expand
View Pricing
BR782841
BRAND
®
insert strips for 24 x 6-well BRANDplates
®
,
Inlet Opening System, PC membrane, pore size 3 μm
Expand
View Pricing
BR782801
BRAND
®
insert strips for 24 x 6-well BRANDplates
®
,
Inlet Opening System, PC membrane, size 0.4 μm
Expand
View Pricing
BR782871
BRAND
®
insert strips for 24 x 6-well BRANDplates
®
,
Inlet Opening System, PET membrane, pore size 8 μm
Expand
View Pricing
BR782811
BRAND
®
insert strips for 24 x 6-well BRANDplates
®
,
Inlet Opening System, PET membrane, pore size 0.4 μm
Expand
View Pricing
BR782861
BRAND
®
insert strips for 24 x 6-well BRANDplates
®
,
Inlet Opening System, PC membrane, pore size 8 μm
Expand
View Pricing