Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
density bottle, Gay-Lussac pattern, uncalibrated
Products
Product Category
bottles (5)
Manufacturer
Capacity (mL)
Cap
Color
Vial Bottle Color
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
BR43208
BRAND
®
density bottle, Gay-Lussac pattern, uncalibrated,
capacity 10 mL, borosilicate glass
Expand
View Pricing
BR43205
BRAND
®
density bottle, Gay-Lussac pattern, uncalibrated,
capacity 5 mL, borosilicate glass
Expand
View Pricing
BR43228
BRAND
®
density bottle, Gay-Lussac pattern, uncalibrated,
capacity 50 mL, borosilicate glass
Expand
View Pricing
BR43220
BRAND
®
density bottle, Gay-Lussac pattern, uncalibrated,
capacity 25 mL, borosilicate glass
Expand
View Pricing
BR43238
BRAND
®
density bottle, Gay-Lussac pattern, uncalibrated,
capacity 100 mL, borosilicate glass
Expand
View Pricing