Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bovine Collagen Type I
Products
Product Category
attachment factors (4)
Brand
Sigma-Aldrich (4)
Biological Source
Gene Alias
Physical Form
Product Line
Tissue
Quality Segment
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
C4243
Bovine Collagen Type I,
2.9-3.2 mg/mL, suitable for cell culture, sterile-filtered
Expand
View Pricing
5162
Bovine Collagen Type I,
from bovine flexor tendon, powder, suitable for cell culture, used for 3D gel formation
Expand
View Pricing
5074
Bovine Collagen Type I,
from bovine skin, liquid, 5 mg/mL, suitable for cell culture, used for 3D Hydrogel formation, PureCol
™
EZ Gel
Expand
View Pricing
C2124
Bovine Collagen Type I,
6 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Expand
View Pricing