Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bovine Calf Serum
Synonym(s):
Calf Serum
Products
Product Category
sera (3)
Brand
Sigma-Aldrich (3)
Biological Source
Chemical Composition
Quality Segment
Geographic Origin
Sterilization Method
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
12133C
Bovine Calf Serum,
for cell culture, USA origin, sterile-filtered, suitable for cell culture
Expand
View Pricing
12138C
Bovine Calf Serum,
Iron-Fortified, USA origin, from formula-fed bovine calves, sterile-filtered, suitable for cell culture
Expand
View Pricing
13067C
Bovine Calf Serum,
Australia origin, γ-irradiated by SER-TAIN
™
process, sterile-filtered, suitable for cell culture
Expand
View Pricing