
(−)-Borneol

Synonym(s):
endo-(1S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing