
Boc-3-(3-pyridyl)-Ala-OH

Synonym(s):
Boc-β-(3-pyridyl)-Ala-OH, Boc-3-(3-pyridyl)-L-alanine
Empirical Formula (Hill Notation):
C13H18N2O4
CAS Number:
Molecular Weight:
266.29
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing