Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
β-Nicotinamide adenine dinucleotide phosphate sodium salt
Synonym(s):
β-NADP-Na, β-NADP-sodium salt, Coenzyme II sodium salt, NADP, TPN, TPN-Na, Triphosphopyridine nucleotide sodium salt
Linear Formula:
C
21
H
27
N
7
O
17
P
3
· Na
CAS Number:
1184-16-3
Molecular Weight:
765.39
EC Number:
214-664-6
Products
Product Category
nucleotides (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
N8035
β-Nicotinamide adenine dinucleotide phosphate sodium salt,
pkg of 5 mg (per vial)
Expand
View Pricing
N8160
β-Nicotinamide adenine dinucleotide phosphate sodium salt,
pkg of 10 mg (per vial)
Expand
View Pricing