
Bluo-Gal

Synonym(s):
5-Bromo-3-indolyl β-D-galactopyranoside, Bluo-Gal
Empirical Formula (Hill Notation):
C14H16BrNO6
CAS Number:
Molecular Weight:
374.18
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Sterility
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing