Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bismuth(III) oxide
Synonym(s):
Dibismuth trioxide, Bismuth(III) oxide
Empirical Formula (Hill Notation):
Bi
2
O
3
CAS Number:
1304-76-3
Molecular Weight:
465.96
EC Number:
215-134-7
Products
Brand
Sigma-Aldrich (4)
Supelco (1)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Particle Size (µm)
Reagent Type
Core
Special Grade
Purum (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
223891
Bismuth(III) oxide,
ReagentPlus
®
, powder, 10 μm, 99.9% trace metals basis
Expand
View Pricing
202827
Bismuth(III) oxide,
powder, 99.999% trace metals basis
Expand
View Pricing
637017
Bismuth(III) oxide,
nanopowder, 90-210 nm particle size, 99.8% trace metals basis
Expand
View Pricing
1.01862
Bismuth(III) oxide,
EMPLURA
®
Expand
View Pricing
95381
Bismuth(III) oxide,
purum, ≥98.0% (KT)
Expand
View Pricing