
Benzyl disulfide

Synonym(s):
Dibenzyl disulfide, Benzyl disulfide, Bis(phenylmethyl) disulfide
Linear Formula:
C6H5CH2SSCH2C6H5
CAS Number:
Molecular Weight:
246.39
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing