Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Benchmark Pulse 150 Horn
Synonym(s):
ultrasonic homogenizer, sonicator, pulsating sonication
Products
Product Category
homogenizers (4)
Manufacturer
Diameter (mm)
Working Volume (ml)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
BMSDP01502
Benchmark Pulse 150 Horn,
diam. 2 mm
Expand
View Pricing
BMSDP01503
Benchmark Pulse 150 Horn,
diam. 3 mm
Expand
View Pricing
BMSDP01506
Benchmark Pulse 150 Horn,
diam. 6 mm, working volume 10100 mL
Expand
View Pricing
BMSDP01508
Benchmark Pulse 150 Horn,
diam. 8 mm, working volume 25150 mL
Expand
View Pricing