
Barium chloride dihydrate

Synonym(s):
Barium chloride dihydrate
Empirical Formula (Hill Notation):
BaCl2 · 2H2O
CAS Number:
Molecular Weight:
244.26
Products
Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Showing 1-7 of 7
Compare
Product Number
Description
Pricing