Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Barbituric acid
Synonym(s):
Barbituric acid, 2,4,6-Trihydroxypyrimidine, Malonylurea
Empirical Formula (Hill Notation):
C
4
H
4
N
2
O
3
CAS Number:
67-52-7
Molecular Weight:
128.09
EC Number:
200-658-0
Products
Product Category
acids (1)
organic acids (1)
Brand
Sigma-Aldrich (2)
Supelco (2)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
1.00132
Barbituric acid,
for analysis EMSURE
®
Expand
View Pricing
185698
Barbituric acid,
ReagentPlus
®
, 99%
Expand
View Pricing
8.00133
Barbituric acid,
for synthesis
Expand
View Pricing
11709
Barbituric acid,
for spectrophotometric det. of cyanide, ≥99.5%
Expand
View Pricing