Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Azelaic acid
Synonym(s):
Nonanedioic acid, Azelaic acid,, Azelaic acid
Linear Formula:
HO
2
C(CH
2
)
7
CO
2
H
CAS Number:
123-99-9
Molecular Weight:
188.22
EC Number:
204-669-1
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (5)
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Monomer Type
Reference Material Type
Special Grade
Technical (2)
analytical standard (1)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
246379
Azelaic acid,
98%
Expand
View Pricing
8.20116
Azelaic acid,
technical grade for synthesis
Expand
View Pricing
A96150
Azelaic acid,
technical grade, 80%
Expand
View Pricing
95054
Azelaic acid,
analytical standard
Expand
View Pricing
8.20117
Azelaic acid,
for synthesis
Expand
View Pricing
11480
Azelaic acid,
technical, ~85% (GC)
Expand
View Pricing