
Ascorbyl Stearate

Synonym(s):
L-Ascorbic acid 6-stearate, 6-O-Stearyl-L-ascorbic acid, L-Ascorbyl 6-octadecanoate, Ascorbyl stearate
Empirical Formula (Hill Notation):
C24H42O7
CAS Number:
Molecular Weight:
442.59
Products
Formula Weight
Manufacturer
Agency
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing