
Aripiprazole Hydroxydihydroquinolinone Analog

Synonym(s):
7-Hydroxy-3,4-dihydro-2(1H)-quinolinone, 7-Hydroxy-1,2,3,4-tetrahydro-2-quinolinone, 7-Hydroxy-3,4-dihydrocarbostyril
Empirical Formula (Hill Notation):
C9H9NO2
CAS Number:
Molecular Weight:
163.17
Products
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing