
Ammonium chloride

Synonym(s):
Salmiac, Ammonium chloride, Ammonium Chloride
Linear Formula:
NH4Cl
Products
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Technique
Showing 1-18 of 18
Compare
Product Number
Description
Pricing