Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonia solution
Linear Formula:
NH
3
CAS Number:
7664-41-7
Molecular Weight:
17.03
Products
Product Category
inorganic bases (4)
salts (3)
Brand
Sigma-Aldrich (7)
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Application
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
499145
Ammonia solution,
7 N in methanol
Expand
View Pricing
407666
Ammonia solution,
0.4 M in dioxane
Expand
View Pricing
392685
Ammonia solution,
2.0 M in ethanol
Expand
View Pricing
779423
Ammonia solution,
4 M in methanol
Expand
View Pricing
718939
Ammonia solution,
0.4 M in THF
Expand
View Pricing
341428
Ammonia solution,
2.0 M in methanol
Expand
View Pricing
392693
Ammonia solution,
2.0 M in isopropanol
Expand
View Pricing