Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum chloride hexahydrate
Empirical Formula (Hill Notation):
AlCl
3
· 6H
2
O
CAS Number:
7784-13-6
Molecular Weight:
241.43
EC Number:
231-208-1
Products
Product Category
acids (2)
Brand
Sigma-Aldrich (2)
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Reagent Type
Core
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
237078
Aluminum chloride hexahydrate,
ReagentPlus
®
, 99%
Expand
View Pricing
A0718
Aluminum chloride hexahydrate,
meets EP, BP, USP testing specifications
Expand
View Pricing