Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum alloy 3003
Products
Manufacturer
Physical Form
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GF11606284
Aluminum alloy 3003,
honeycomb, Al 98%/Mn 1%/Fe 0.5%/Si 0.5%, thickness 100 mm, size 300 × 300 mm, cell size 6.35 mm
Expand
View Pricing
GF10292688
Aluminum alloy 3003,
honeycomb, Al 98%/Mn 1%/Fe 0.5%/Si 0.5%, thickness 20 mm, size 600 × 600 mm, cell size 12.7 mm
Expand
View Pricing
GF47951815
Aluminum alloy 3003,
honeycomb, Al 98%/Mn 1%/Fe 0.5%/Si 0.5%, thickness 100 mm, size 600 × 600 mm, cell size 6.35 mm
Expand
View Pricing
GF64165057
Aluminum alloy 3003,
honeycomb, Al98%/Mn 1%/Fe 0.5%/Si 0.5%, thickness 20 mm, size 300 × 300 mm, cell size 6.35 mm
Expand
View Pricing
GF84866169
Aluminum alloy 3003,
honeycomb, thickness 20 mm, Al98%/Mn 1%/Fe 0.5%/Si 0.5%, size 300 × 300 mm, cell size 12.7 mm
Expand
View Pricing
GF91010854
Aluminum alloy 3003,
honeycomb, thickness 20 mm, Al98%/Mn 1%/Fe 0.5%/Si 0.5%, size 600 × 600 mm, cell size 6.35 mm
Expand
View Pricing