Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
Sure/Stor
™
flask
Products
Product Category
flasks (12)
Brand
Sigma-Aldrich (12)
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Style
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
Z404993
Aldrich
®
Sure/Stor
™
flask,
capacity 100 mL, clear glass
Expand
View Pricing
Z405027
Aldrich
®
Sure/Stor
™
flask,
capacity 1,000 mL, clear glass
Expand
View Pricing
Z405000
Aldrich
®
Sure/Stor
™
flask,
capacity 250 mL, clear glass
Expand
View Pricing
Z404977
Aldrich
®
Sure/Stor
™
flask,
capacity 25 mL, clear glass
Expand
View Pricing
Z404985
Aldrich
®
Sure/Stor
™
flask,
capacity 50 mL, clear glass
Expand
View Pricing
Z405116
Aldrich
®
Sure/Stor
™
flask,
capacity 2,000 mL, plastic-coated glass
Expand
View Pricing
Z405019
Aldrich
®
Sure/Stor
™
flask,
capacity 500 mL, clear glass
Expand
View Pricing
Z405108
Aldrich
®
Sure/Stor
™
flask,
capacity 1,000 mL, plastic-coated glass
Expand
View Pricing
Z405078
Aldrich
®
Sure/Stor
™
flask,
capacity 100 mL, plastic-coated glass
Expand
View Pricing
Z405043
Aldrich
®
Sure/Stor
™
flask,
capacity 25 mL, plastic-coated glass
Expand
View Pricing
Z405051
Aldrich
®
Sure/Stor
™
flask,
capacity 50 mL, plastic-coated glass
Expand
View Pricing
Z405094
Aldrich
®
Sure/Stor
™
flask,
capacity 500 mL, plastic-coated glass
Expand
View Pricing