Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
super condenser
Products
Brand
Sigma-Aldrich (6)
Height (mm)
OD (mm)
Surface Area cm²
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z224979
Aldrich
®
super condenser,
Overall H 320 mm, joint: ST/NS 24/40
Expand
View Pricing
Z225002
Aldrich
®
super condenser,
Overall H 510 mm, joint: ST/NS 24/40
Expand
View Pricing
Z517437
Aldrich
®
super condenser,
Overall H 510 mm, joint: ST/NS 29/32
Expand
View Pricing
Z517429
Aldrich
®
super condenser,
Overall H 410 mm, joint: ST/NS 29/32
Expand
View Pricing
Z224987
Aldrich
®
super condenser,
Overall H 370 mm, joint: ST/NS 24/40
Expand
View Pricing
Z224995
Aldrich
®
super condenser,
Overall H 410 mm, joint: ST/NS 24/40
Expand
View Pricing