Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
solvent-pouring funnel
Products
Product Category
funnels (5)
Brand
Sigma-Aldrich (5)
Length (mm)
OD (mm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z548790
Aldrich
®
solvent-pouring funnel,
stem L × O.D. 65 mm × 15 mm
Expand
View Pricing
Z548839
Aldrich
®
solvent-pouring funnel,
stem L × O.D. 100 mm × 28 mm
Expand
View Pricing
Z548820
Aldrich
®
solvent-pouring funnel,
stem L × O.D. 100 mm × 22 mm
Expand
View Pricing
Z548804
Aldrich
®
solvent-pouring funnel,
stem L × O.D. 65 mm × 22 mm
Expand
View Pricing
Z548812
Aldrich
®
solvent-pouring funnel,
stem L × O.D. 65 mm × 28 mm
Expand
View Pricing