Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Alcohol Dehydrogenase from
Saccharomyces cerevisiae
Synonym(s):
ADH, Alcohol Dehydrogenase from yeast, Alcohol:NAD
+
oxidoreductase
CAS Number:
9031-72-5
EC Number:
232-870-4
Products
Product Category
enzymes (3)
Brand
Sigma-Aldrich (3)
Sustainability
Biological Source
Color
Feature
Physical Form
Quality Segment
Application
Package Size
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A7011
Alcohol Dehydrogenase from
Saccharomyces cerevisiae
,
≥300 units/mg protein, lyophilized powder (contains buffer salts), M
w
141-151 kDa
Expand
View Pricing
A3263
Alcohol Dehydrogenase from
Saccharomyces cerevisiae
,
powder, ≥300 units/mg protein, mol wt ~141,000 (four subunits)
Expand
View Pricing
A8656
Alcohol Dehydrogenase from
Saccharomyces cerevisiae
,
Expand
View Pricing