Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Al alloy
Synonym(s):
Aluminum alloy
Products
Manufacturer
Agency
Format
Reference Material Type
Special Grade
certified reference material (7)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
IRMM532A
Al alloy,
IRMM
®
, certified reference material, 0.01% Co, 0.1 mm foil
Expand
View Pricing
ERMEB530B
Al alloy,
0.1% Au, 0.5 mm wire, ERM
®
, certified reference material
Expand
View Pricing
ERMEB530C
Al alloy,
0.1% Au, 1.0 mm wire, ERM
®
, certified reference material
Expand
View Pricing
IRMM534B
Al alloy,
IRMM
®
, certified reference material, 2.0% Sc, 0.5 mm wire
Expand
View Pricing
IRMM534A
Al alloy,
IRMM
®
, certified reference material, 2.0% Sc, 0.1 mm foil
Expand
View Pricing
IRMM532C
Al alloy,
IRMM
®
, certified reference material, 0.01% Co, 1.0 mm wire
Expand
View Pricing
IRMM532B
Al alloy,
IRMM
®
, certified reference material, 0.01% Co, 0.5 mm wire
Expand
View Pricing