Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Agarose, low gelling temperature
Synonym(s):
2-Hydroxyethyl agarose
CAS Number:
39346-81-1
Products
Product Category
carbohydrates (4)
Biological Source
Feature
Melting Point (°C)
Physical Form
Product Line
Research Area
Quality Segment
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
A9414
Agarose, low gelling temperature,
BioReagent, Molecular Biology
Expand
View Pricing
A9045
Agarose, low gelling temperature,
BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Expand
View Pricing
A4018
Agarose, low gelling temperature,
Expand
View Pricing
A0701
Agarose, low gelling temperature,
Type VII-A
Expand
View Pricing