Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adapter for Eppendorf
®
S-4-72 Rotor
Products
Product Category
adapters (9)
Manufacturer
Diameter (mm)
Height (mm)
Package Size
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
EP5804787005
Adapter for Eppendorf
®
S-4-72 Rotor,
capacity, 1 x 250 mL flat/175 - 225 mL bottle, pack of 2
Expand
View Pricing
EP5804794001
Adapter for Eppendorf
®
S-4-72 Rotor,
holds 26 x 1.5/2 mL tubes, pack of 2 ea
Expand
View Pricing
EP5804783000
Adapter for Eppendorf
®
S-4-72 Rotor,
holds 8 x 15 mL conical tubes, pack of 2
Expand
View Pricing
EP5804784006
Adapter for Eppendorf
®
S-4-72 Rotor,
holds 4 x 50 mL conical tubes, pack of 2 ea
Expand
View Pricing
EP5804789008
Adapter for Eppendorf
®
S-4-72 Rotor,
holds 14 x 2.6-8 mL tubes, pack of 2 ea
Expand
View Pricing
EP5804785002
Adapter for Eppendorf
®
S-4-72 Rotor,
holds 2 x 50 mL conical tubes, pack of 2 ea
Expand
View Pricing
EP5804791002
Adapter for Eppendorf
®
S-4-72 Rotor,
holds 13 x 5.5 - 12 mL round-bottom tubes, pack of 2 ea
Expand
View Pricing
EP5804792009
Adapter for Eppendorf
®
S-4-72 Rotor,
holds 12 x 9 mL round-bottom tubes, pack of 2 ea
Expand
View Pricing
EP5804793005
Adapter for Eppendorf
®
S-4-72 Rotor,
holds 8 x Eppendorf Tube
®
5.0 mL, pack of 2 ea
Expand
View Pricing