Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adapter for Eppendorf
®
A-4-81 Rotor
Products
Product Category
adapters (11)
Manufacturer
Diameter (mm)
Height (mm)
Package Size
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
ADAPA481550-EP
Adapter for Eppendorf
®
A-4-81 Rotor,
holds 5 x 50 mL conical tubes, for 500 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA481180-EP
Adapter for Eppendorf
®
A-4-81 Rotor,
holds 1 bottle 180 - 250 mL flat/175 - 225 mL conical
Expand
View Pricing
ADAPA481125-EP
Adapter for Eppendorf
®
A-4-81 Rotor,
holds 12 Eppendorf Tube
®
5.0 mL and 15 mL conical tubes 15 mL, for 500 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA481201-EP
Adapter for Eppendorf
®
A-4-81 Rotor,
holds 20 x 1.2 - 5 mL reaction vessels, for 500 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA4812015-EP
Adapter for Eppendorf
®
A-4-81 Rotor,
holds 20 x 1.5 - 2.0 mL reaction vessels, for 500 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA481252-EP
Adapter for Eppendorf
®
A-4-81 Rotor,
holds 25 x 2.6–5 mL round-bottom tubes, for 500 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA481163-EP
Adapter for Eppendorf
®
A-4-81 Rotor,
holds 16 x 3 - 15 mL round-bottom tubes, for 500 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA4815SC-EP
Adapter for Eppendorf
®
A-4-81 Rotor,
holds 5 conical tubes, skirted/Centriprep, for 500 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA481167-EP
Adapter for Eppendorf
®
A-4-81 Rotor,
holds 16 x 7 - 17 mL round-bottom tubes, for 500 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA481140-EP
Adapter for Eppendorf
®
A-4-81 Rotor,
holds 1 x 400 mL bottle, for 500 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA481182-EP
Adapter for Eppendorf
®
A-4-81 Rotor,
holds 18 x 2.6 -7 mL reaction vessels, for 500 mL rectangular buckets, pack of 2 ea
Expand
View Pricing