Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adapter for Eppendorf
®
A-4-62 Rotor
Products
Product Category
adapters (14)
Manufacturer
Diameter (mm)
Height (mm)
Package Size
Technique
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
ADAPA462915-EP
Adapter for Eppendorf
®
A-4-62 Rotor,
holds 9 x 15 mL conical tubes, for 250 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA462350-EP
Adapter for Eppendorf
®
A-4-62 Rotor,
holds 3 x 50 mL conical tubes, for 250 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA462200-EP
Adapter for Eppendorf
®
A-4-62 Rotor,
holds 1 x 200 - 250 mL flat bottle / 175 - 225 mL conical bottle, for 250 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA462450C-EP
Adapter for Eppendorf
®
A-4-62 Rotor,
holds conical tubes, for 250 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA462255-EP
Adapter for Eppendorf
®
A-4-62 Rotor,
holds 25 x 1.2–5 mL blood tubes, for 250 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA4621620-EP
Adapter for Eppendorf
®
A-4-62 Rotor,
holds 16 x 1.5 - 2.0 mL reaction vessels, for 250 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA462157-EP
Adapter for Eppendorf
®
A-4-62 Rotor,
holds 15 x 2.6 - 7 mL blood tubes, for 250 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA462430-EP
Adapter for Eppendorf
®
A-4-62 Rotor,
holds mL round-bottom tubes, for 250 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA462120-EP
Adapter for Eppendorf
®
A-4-62 Rotor,
holds 1 x 80 - 120 mL tubes, for 250 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA462520-EP
Adapter for Eppendorf
®
A-4-62 Rotor,
holds 2 x 50 - 75 mL round-bottom tubes, for 250 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA462450-EP
Adapter for Eppendorf
®
A-4-62 Rotor,
holds 4 x 30 - 50 mL conical tubes, for 250 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA462818-EP
Adapter for Eppendorf
®
A-4-62 Rotor,
holds 8 x 7 - 18 mL round-bottom tubes, for 250 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA4621217-EP
Adapter for Eppendorf
®
A-4-62 Rotor,
holds 12 x 7–17 mL blood tubes, for 250 mL rectangular buckets, pack of 2 ea
Expand
View Pricing
ADAPA4621215-EP
Adapter for Eppendorf
®
A-4-62 Rotor,
holds 12 x 3–15 mL blood tubes, for 250 mL rectangular buckets, pack of 2 ea
Expand
View Pricing