Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ace Trubore
®
stirrer bearing
Products
Brand
Sigma-Aldrich (2)
Manufacturer
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z125911
Ace Trubore
®
stirrer bearing,
fits shaft o.d., 10 mm, joint: ST/NS 24/40
Expand
View Pricing
Z125954
Ace Trubore
®
stirrer bearing,
joint: ST/NS 24/40, fits shaft o.d., 10 mm
Expand
View Pricing
Z125962
Ace Trubore
®
stirrer bearing,
joint: ST/NS 29/42, fits shaft o.d., 10 mm
Expand
View Pricing
Z154601
Ace Trubore
®
stirrer bearing,
fits shaft o.d., 6 mm, joint: ST/NS 14/20
Expand
View Pricing
Z154598
Ace Trubore
®
stirrer bearing,
fits shaft o.d., 5 mm, joint: ST/NS 14/20
Expand
View Pricing
Z154628
Ace Trubore
®
stirrer bearing,
joint: ST/NS 19/22, fits shaft o.d., 6 mm
Expand
View Pricing
Z154288
Ace Trubore
®
stirrer bearing,
joint: ST/NS 19/22, fits shaft o.d., 10 mm
Expand
View Pricing
Z154296
Ace Trubore
®
stirrer bearing,
fits shaft o.d., 10 mm, joint: ST/NS 45/50
Expand
View Pricing
Z125938
Ace Trubore
®
stirrer bearing,
fits shaft o.d., 10 mm, joint: ST/NS 29/42
Expand
View Pricing