Skip to Content
Merck
Acetic anhydride derivatization grade (GC derivatization), LiChropur™, ≥99.0%

Acetic anhydride

Acetic anhydride derivatization grade (GC derivatization), LiChropur™, ≥99.0%
Synonym(s):
Acetic anhydride
Linear Formula:
(CH3CO)2O
CAS Number:
Molecular Weight:
102.09
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade