
Acetic acid (glacial)

Synonym(s):
Acetic acid, Glacial acetic acid
Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing