Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
α-Amylase from
Bacillus licheniformis
Synonym(s):
1,4-α-D-Glucan-glucanohydrolase
CAS Number:
9000-85-5
EC Number:
232-560-9
Products
Brand
Sigma-Aldrich (2)
Supelco (1)
Sustainability
Biological Source
Feature
Physical Form
Purity
Analyte Suitability
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A3403
α-Amylase from
Bacillus licheniformis
,
Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Expand
View Pricing
A4551
α-Amylase from
Bacillus licheniformis
,
lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Expand
View Pricing
A4582
α-Amylase from
Bacillus licheniformis
,
suitable for determination of starch (Kit STA-20)
Expand
View Pricing