Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4-
tert
-Butylcatechol
Linear Formula:
(CH
3
)
3
CC
6
H
3
-1,2-(OH)
2
CAS Number:
98-29-3
Molecular Weight:
166.22
EC Number:
202-653-9
Products
Product Category
building blocks (3)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Puriss (1)
Purum (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
19671
4-
tert
-Butylcatechol,
≥99.0%
Expand
View Pricing
19670
4-
tert
-Butylcatechol,
≥97.0% (HPLC)
Expand
View Pricing
03-4881
4-
tert
-Butylcatechol,
SAJ first grade, ≥98.0%
Expand
View Pricing