Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4-Nitrophenyl dodecanoate
Synonym(s):
4-Nitrophenyl laurate, Dodecanoic acid 4-nitrophenyl ester
Empirical Formula (Hill Notation):
C
18
H
27
NO
4
CAS Number:
1956-11-2
Molecular Weight:
321.41
EC Number:
217-796-2
Products
Product Category
substrates (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
Purity
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
61716
4-Nitrophenyl dodecanoate,
≥98.0% (GC)
Expand
View Pricing