Skip to Content
Merck
4-(Dimethylamino)azobenzene-4′-sulfonyl chloride derivatization grade (HPLC), LiChropur™, 98.0-102.0% (AT)

4-(Dimethylamino)azobenzene-4′-sulfonyl chloride

4-(Dimethylamino)azobenzene-4′-sulfonyl chloride derivatization grade (HPLC), LiChropur™, 98.0-102.0% (AT)
Synonym(s):
4-(4-Dimethylaminophenylazo)benzenesulfonyl chloride, DABS-Cl, Dabsyl chloride
Linear Formula:
(CH3)2NC6H4N=NC6H4SO2Cl
CAS Number:
Molecular Weight:
323.80
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Technique