
4-Amino-2,2,6,6-tetramethylpiperidine

Synonym(s):
Triacetonediamine, 4-Amino-2,2,6,6-tetramethylpiperidine
Empirical Formula (Hill Notation):
C9H20N2
CAS Number:
Molecular Weight:
156.27
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing