
4-(4-Methylphenyl)-4-oxobutyric acid

Synonym(s):
3-p-Toluoylpropionic acid
Linear Formula:
CH3C6H4COCH2CH2CO2H
CAS Number:
Molecular Weight:
192.21
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing