Skip to Content
Merck
3-Pyridineboronic acid pinacol ester 97%

3-Pyridineboronic acid pinacol ester

3-Pyridineboronic acid pinacol ester 97%
Synonym(s):
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Pyridylboronic acid pinacol ester
Empirical Formula (Hill Notation):
C11H16BNO2
CAS Number:
Molecular Weight:
205.06

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing