Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-(Methylthio)ethanol
Synonym(s):
2-(Methylmercapto)ethanol, 2-Hydroxyethyl methyl sulfide
Linear Formula:
CH
3
SCH
2
CH
2
OH
CAS Number:
5271-38-5
Molecular Weight:
92.16
EC Number:
226-090-3
Products
Product Category
aroma chemicals (1)
building blocks (1)
Brand
Sigma-Aldrich (2)
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
226424
2-(Methylthio)ethanol,
99%
Expand
View Pricing
W400408
2-(Methylthio)ethanol,
≥99%
Expand
View Pricing