
2-Methylfuran

Synonym(s):
Silvan, 2-Methylfuran
Empirical Formula (Hill Notation):
C5H6O
CAS Number:
Molecular Weight:
82.10
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing